siRNA Details
| Gene Symbol: AA415398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004178.1 | NP_001004178.1 | 4 | 1224 | ||
| GenBank mRNA | BC064115.1 | - | 1 | 490 | 3417656 | |
| BC076608.1 | - | 4 | 1224 | 30625085 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AA415398| Gene Symbol: AA415398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004178.1 | NP_001004178.1 | 4 | 1224 | ||
| GenBank mRNA | BC064115.1 | - | 1 | 490 | 3417656 | |
| BC076608.1 | - | 4 | 1224 | 30625085 | ||