siRNA Details
| Gene Symbol: Trim63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039048.2 | NP_001034137.2 | 1 | 189 | ||
| GenBank mRNA | AK052911.1 | - | 1 | 100 | ||
| BC147663.1 | - | 1 | 157 | 9087452 | ||
| BC147666.1 | - | 1 | 157 | 9087455 | ||
| CJ131783.1 | - | 1 | 191 | |||
| CJ131880.1 | - | 1 | 190 | |||
| DQ229108.1 | - | 1 | 85 | |||

Trim63
Genomic Map