siRNA Details
| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163006.2 | NP_001156478.1 | 2, 3 | 235 | ||
| GenBank mRNA | AA023452.1 | - | 2, 3 | 250 | ||
| AK004466.1 | - | 2, 3 | 127 | |||
| AK008362.1 | - | 2, 3 | 119 | |||
| AK008449.1 | - | 2, 3 | 119 | |||
| AK009189.1 | - | 2, 3 | 118 | |||
| AK009528.1 | - | 2, 3 | 161 | |||
| AK011553.1 | - | 2, 3 | 118 | |||
| AK030343.1 | - | 2, 3 | 188 | |||
| AK152509.1 | - | 2, 3 | 161 | |||
| AK153063.1 | - | 2, 3 | 160 | |||
| AK159039.1 | - | 2, 3 | 114 | |||
| AK159100.1 | - | 2, 3 | 161 | |||
| AK167818.1 | - | 2, 3 | 160 | |||
| AW226897.1 | - | 2, 3 | 293 | |||
| BC055791.1 | - | 2, 3 | 152 | 6410794 | ||
| BY776411.1 | - | 2, 3 | 225 | |||

Minos1
Genomic Map