siRNA Details
| Gene Symbol: Gm13119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034101.2 | NP_001029273.2 | 4 | 1378 | ||
| GenBank mRNA | BC100406.1 | - | 3 | 1378 | 30727972 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13119| Gene Symbol: Gm13119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034101.2 | NP_001029273.2 | 4 | 1378 | ||
| GenBank mRNA | BC100406.1 | - | 3 | 1378 | 30727972 | |