siRNA Details
| Gene Symbol: Rnf207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033489.2 | NP_001028661.2 | 13 | 1603 | ||
| GenBank mRNA | AK042379.1 | - | 13 | 1586 | ||
| BC141256.1 | - | 13 | 1487 | 9055868 | ||
| BC141257.1 | - | 13 | 1487 | 9055869 | ||

Rnf207
Genomic Map