siRNA Details
| Gene Symbol: AI506816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK151523.1 | - | 4 | 1034 | ||
| AK161191.1 | - | 4 | 1044 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI506816| Gene Symbol: AI506816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK151523.1 | - | 4 | 1034 | ||
| AK161191.1 | - | 4 | 1044 | |||