siRNA Details
| Gene Symbol: Ociad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026950.3 | NP_081226.1 | 3 | 299 | ||
| GenBank mRNA | AK007617.1 | - | 3 | 253 | ||
| BC044883.1 | - | 3 | 218 | 4971755 | ||
| BY250863.1 | - | 3 | 328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ociad2| Gene Symbol: Ociad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026950.3 | NP_081226.1 | 3 | 299 | ||
| GenBank mRNA | AK007617.1 | - | 3 | 253 | ||
| BC044883.1 | - | 3 | 218 | 4971755 | ||
| BY250863.1 | - | 3 | 328 | |||