siRNA Details
Gene Symbol: Gm3286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122678.2 | NP_001116150.1 | 4 | 1898 | ||
NM_001167792.1 | NP_001161264.1 | 3 | 1307 | |||
NM_001167793.1 | NP_001161265.1 | 4 | 1938 | |||
GenBank mRNA | AK145737.1 | - | 6 | 2194 |
Gene Symbol: D5Ertd577e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177187.4 | NP_796161.3 | 4 | 1900 | ||
GenBank mRNA | AK054417.1 | - | 4 | 1900 | ||
BC053383.1 | - | 3 | 1809 | |||
BC094905.1 | - | 3 | 1797 | 30047197 | ||
BC099941.1 | - | 4 | 1884 | 30031929 |
Gene Symbol: E330014E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122668.1 | NP_001116140.1 | 4 | 2020 | ||
GenBank mRNA | AK087741.1 | - | 4 | 2023 | ||
BB709961.1 | - | 1 | 315 |
