siRNA Details
| Gene Symbol: Mn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081235.1 | NP_001074704.1 | 1 | 3103 | ||
| GenBank mRNA | AK142207.1 | - | 2214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mn1| Gene Symbol: Mn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081235.1 | NP_001074704.1 | 1 | 3103 | ||
| GenBank mRNA | AK142207.1 | - | 2214 | |||