siRNA Details
| Gene Symbol: Tmem178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004182.3 | NP_001004182.1 | 3 | 314 | ||
| GenBank mRNA | AK084030.1 | - | 3 | 315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem178b| Gene Symbol: Tmem178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004182.3 | NP_001004182.1 | 3 | 314 | ||
| GenBank mRNA | AK084030.1 | - | 3 | 315 | ||