siRNA Details
| Gene Symbol: Vmn1r20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101533.1 | NP_001095003.1 | 1 | 871 | ||
| Gene Symbol: Vmn1r27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134436.2 | NP_602318.2 | 1 | 871 | ||

Vmn1r20
Genomic Map