siRNA Details
| Gene Symbol: Igkv9-120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U88675.1 | - | 191 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AY498737.1 | - | 89 | Chr. - - [-] on Build | ||

Igkv9-120
Genomic Map