siRNA Details
Gene Symbol: Gm5595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008427.1 | NP_001008427.1 | 4 | 1425 | ||
GenBank mRNA | AB010373.1 | - | 1 | 141 | ||
BC068175.1 | - | 4 | 1425 | 30550329 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008427.1 | NP_001008427.1 | 4 | 1425 | ||
GenBank mRNA | AB010373.1 | - | 1 | 141 | ||
BC068175.1 | - | 4 | 1425 | 30550329 |