siRNA Details
| Gene Symbol: Gm5595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008427.1 | NP_001008427.1 | 4 | 867 | ||
| GenBank mRNA | BC068175.1 | - | 4 | 867 | 30550329 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5595| Gene Symbol: Gm5595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008427.1 | NP_001008427.1 | 4 | 867 | ||
| GenBank mRNA | BC068175.1 | - | 4 | 867 | 30550329 | |