siRNA Details
Gene Symbol: Slc28a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004184.3 | NP_001004184.2 | 15 | 1692 | ||
GenBank mRNA | BC061230.1 | - | 15 | 1692 | 30379959 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc28a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004184.3 | NP_001004184.2 | 15 | 1692 | ||
GenBank mRNA | BC061230.1 | - | 15 | 1692 | 30379959 |