siRNA Details
| Gene Symbol: Slc28a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004184.3 | NP_001004184.2 | 4, 5 | 366 | ||
| GenBank mRNA | BC061230.1 | - | 4, 5 | 366 | 30379959 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc28a1| Gene Symbol: Slc28a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004184.3 | NP_001004184.2 | 4, 5 | 366 | ||
| GenBank mRNA | BC061230.1 | - | 4, 5 | 366 | 30379959 | |