siRNA Details
| Gene Symbol: Lrrc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113379.1 | NP_001106850.1 | 2 | 1141 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc32| Gene Symbol: Lrrc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113379.1 | NP_001106850.1 | 2 | 1141 | ||