siRNA Details
| Gene Symbol: Gm1966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277179.1 | NP_001264108.1 | 3 | 1422 | ||
| NM_001277180.1 | NP_001264109.1 | 3 | 1434 | |||
| NM_001277181.1 | NP_001264110.1 | 4 | 1506 | |||
| NM_001277182.1 | NP_001264111.1 | 4 | 1572 | |||
| NM_001277183.1 | NP_001264112.1 | 6 | 1706 | |||
| GenBank mRNA | AK156879.1 | - | 4 | 1488 | ||
| AK171325.1 | - | 3 | 1436 | |||
| AK172128.1 | - | 6 | 1684 | |||

Gm1966
Genomic Map