siRNA Details
Gene Symbol: Gm5615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033783.2 | NP_001028955.1 | 2 | 185 | ||
GenBank mRNA | AK136744.1 | - | 2 | 186 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033783.2 | NP_001028955.1 | 2 | 185 | ||
GenBank mRNA | AK136744.1 | - | 2 | 186 |