siRNA Details
| Gene Symbol: Gm5617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004191.3 | NP_001004191.2 | 1 | 478 | ||
| GenBank mRNA | AK007723.1 | - | 1 | 383 | ||
| BC011344.1 | - | 1 | 373 | 3481870 | ||
| CA463871.1 | - | 481 | ||||
| CN838422.1 | - | 1 | 382 | |||

Gm5617
Genomic Map