siRNA Details
| Gene Symbol: Fbxw20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008428.3 | NP_001008428.1 | 8 | 1067 | ||
| GenBank mRNA | AK135785.1 | - | 8 | 1048 | ||
| AK136200.1 | - | 8 | 1045 | |||
| BC085247.1 | - | 8 | 1048 | 30477089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw20| Gene Symbol: Fbxw20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008428.3 | NP_001008428.1 | 8 | 1067 | ||
| GenBank mRNA | AK135785.1 | - | 8 | 1048 | ||
| AK136200.1 | - | 8 | 1045 | |||
| BC085247.1 | - | 8 | 1048 | 30477089 | ||