siRNA Details
Gene Symbol: RFXAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000538.3 | NP_000529.1 | 2 | 781 | ||
GenBank mRNA | AK313912.1 | - | 2 | 763 | ||
BC026088.1 | - | 2 | 760 | 4827378 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RFXAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000538.3 | NP_000529.1 | 2 | 781 | ||
GenBank mRNA | AK313912.1 | - | 2 | 763 | ||
BC026088.1 | - | 2 | 760 | 4827378 |