siRNA Details
| Gene Symbol: Slc22a28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013820.3 | NP_001013842.2 | 3, 4 | 860 | ||
| GenBank mRNA | AK052719.1 | - | 3, 4 | 861 | ||
| BC139174.1 | - | 3, 4 | 792 | 8862196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a28| Gene Symbol: Slc22a28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013820.3 | NP_001013842.2 | 3, 4 | 860 | ||
| GenBank mRNA | AK052719.1 | - | 3, 4 | 861 | ||
| BC139174.1 | - | 3, 4 | 792 | 8862196 | ||