siRNA Details
| Gene Symbol: Scgb1b19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281526.1 | NP_001268455.1 | 2 | 169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1b19| Gene Symbol: Scgb1b19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281526.1 | NP_001268455.1 | 2 | 169 | ||