siRNA Details
| Gene Symbol: Rhox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024850.2 | NP_001020021.1 | 1 | 406 | ||
| GenBank mRNA | AK146090.1 | - | 1 | 388 | ||
| BC100366.1 | - | 1 | 350 | 30524902 | ||
| BC132474.1 | - | 1 | 337 | 40130751 | ||
| BC132500.1 | - | 1 | 337 | 40130777 | ||
| DQ058647.1 | - | 1 | 406 | |||

Rhox10
Genomic Map