siRNA Details
| Gene Symbol: Ccdc160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034059.1 | NP_001029231.1 | 2 | 789 | ||
| GenBank mRNA | AK134708.1 | - | 2 | 789 | ||
| BC138946.1 | - | 2 | 737 | 8861968 | ||
| BC138947.1 | - | 2 | 737 | 8861969 | ||

Ccdc160
Genomic Map