siRNA Details
| Gene Symbol: Mageb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033492.2 | NP_001028664.2 | 1263 | |||
| GenBank mRNA | AK144342.1 | - | 1264 | |||
| AK144350.1 | - | 1229 | ||||
| AK165797.1 | - | 1424 | ||||
| BC150912.1 | - | 1208 | 9087823 | |||
| BC172073.1 | - | 1208 | 9054359 | |||

Mageb4
Genomic Map