siRNA Details
| Gene Symbol: Gm21943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017393.3 | NP_001017393.1 | 2 | 981 | ||
| GenBank mRNA | BC089465.1 | - | 2 | 981 | 6707203 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm21943| Gene Symbol: Gm21943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017393.3 | NP_001017393.1 | 2 | 981 | ||
| GenBank mRNA | BC089465.1 | - | 2 | 981 | 6707203 | |