siRNA Details
Gene Symbol: Gm21943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017393.3 | NP_001017393.1 | 2 | 1167 | ||
GenBank mRNA | BC089465.1 | - | 2 | 1167 | 6707203 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm21943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017393.3 | NP_001017393.1 | 2 | 1167 | ||
GenBank mRNA | BC089465.1 | - | 2 | 1167 | 6707203 |