siRNA Details
Gene Symbol: Glyatl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145060.1 | NP_001138532.1 | 3 | 202 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
