siRNA Details
Gene Symbol: Gm13177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081248.1 | NP_001074717.1 | 4 | 967 | ||
GenBank mRNA | BC150982.1 | - | 4 | 1016 | 9087895 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm13177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081248.1 | NP_001074717.1 | 4 | 967 | ||
GenBank mRNA | BC150982.1 | - | 4 | 1016 | 9087895 |