siRNA Details
| Gene Symbol: Zfp936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034893.1 | NP_001030065.1 | 4 | 899 | ||
| GenBank mRNA | AK145615.1 | - | 4 | 899 | ||
| AK161338.1 | - | 846 | ||||
| BC066874.1 | - | 1 | 829 | 30106557 | ||
| Gene Symbol: Ckap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132136.1 | - | 2540 | |||

Zfp936
Genomic Map