siRNA Details
| Gene Symbol: Fam92b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033980.2 | NP_001029152.2 | 3 | 438 | ||
| GenBank mRNA | AK131790.1 | - | 3 | 438 | ||
| BC147233.1 | - | 3 | 357 | 9056283 | ||
| BC147234.1 | - | 3 | 357 | 9056269 | ||
| BC172014.1 | - | 2 | 122 | |||

Fam92b
Genomic Map