siRNA Details
| Gene Symbol: Gpr62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159652.1 | NP_001153124.1 | 1 | 1615 | ||
| GenBank mRNA | AK005968.1 | - | 1 | 606 | ||
| AK015060.1 | - | 1 | 593 | |||
| AK015270.1 | - | 1 | 602 | |||
| AK016613.1 | - | 1 | 593 | |||
More Information
Set Membership: |
|

Gpr62
Genomic Map