siRNA Details
| Gene Symbol: Gpr62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159652.1 | NP_001153124.1 | 1 | 363 | ||
| GenBank mRNA | AY255551.1 | - | 1 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr62| Gene Symbol: Gpr62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159652.1 | NP_001153124.1 | 1 | 363 | ||
| GenBank mRNA | AY255551.1 | - | 1 | 102 | ||
Set Membership: |
|