siRNA Details
| Gene Symbol: Btg1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243943.1 | NP_001230872.1 | 5 | 771 | ||
| GenBank mRNA | AK132937.1 | - | 5 | 772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btg1-ps1| Gene Symbol: Btg1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243943.1 | NP_001230872.1 | 5 | 771 | ||
| GenBank mRNA | AK132937.1 | - | 5 | 772 | ||