siRNA Details
| Gene Symbol: Rab44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002786.2 | NP_001002786.1 | 4, 5 | 3017 | ||
| GenBank mRNA | AK036563.1 | - | 4, 5 | 3018 | ||
| BC139372.1 | - | 4, 5 | 2625 | 8862394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab44| Gene Symbol: Rab44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002786.2 | NP_001002786.1 | 4, 5 | 3017 | ||
| GenBank mRNA | AK036563.1 | - | 4, 5 | 3018 | ||
| BC139372.1 | - | 4, 5 | 2625 | 8862394 | ||