siRNA Details
| Gene Symbol: RHEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005614.3 | NP_005605.1 | 8 | 892 | ||
| XM_011516457.1 | XP_011514759.1 | 9 | 946 | |||
| GenBank mRNA | AF493921.1 | - | 8 | 479 | ||
| AK125446.1 | - | 8 | 890 | |||
| AK289370.1 | - | 8 | 621 | |||
| BC016155.1 | - | 8 | 892 | 3528583 | ||
| BC066307.1 | - | 1 | 650 | 5274556 | ||
| BC107705.1 | - | 1 | 841 | 6615350 | ||
| BT006958.1 | - | 8 | 479 | |||
| D78132.1 | - | 8 | 496 | |||
| Z29677.1 | - | 8 | 502 | |||

RHEB
Genomic Map