siRNA Details
| Gene Symbol: Xrra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164258.1 | NP_001157730.1 | 13 | 1379 | ||
| GenBank mRNA | AK154558.1 | - | 14 | 1482 | ||
| BC151014.1 | - | 13 | 1379 | 9087927 | ||
| BC151020.1 | - | 13 | 1379 | 9087933 | ||

Xrra1
Genomic Map