siRNA Details
| Gene Symbol: Zfp97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011765.5 | NP_035895.3 | 4 | 1373 | ||
| GenBank mRNA | L20450.1 | - | 1341 | |||
| Gene Symbol: Zfp960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC018101.1 | - | 4 | 1228 | 3494875 | |

Zfp97
Genomic Map