siRNA Details
| Gene Symbol: Clec4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005860.2 | NP_001005860.1 | 4 | 361 | ||
| GenBank mRNA | AY397673.1 | - | 4 | 361 | ||
| BC104370.1 | - | 4 | 361 | 40049888 | ||
| BC104371.2 | - | 4 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4a4| Gene Symbol: Clec4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005860.2 | NP_001005860.1 | 4 | 361 | ||
| GenBank mRNA | AY397673.1 | - | 4 | 361 | ||
| BC104370.1 | - | 4 | 361 | 40049888 | ||
| BC104371.2 | - | 4 | 360 | |||