siRNA Details
| Gene Symbol: AY761185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012640.2 | NP_001012658.1 | 1 | 129 | ||
| GenBank mRNA | AY761185.1 | - | 1 | 91 | ||
| BC141303.1 | - | 1 | 129 | 9055915 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AY761185| Gene Symbol: AY761185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012640.2 | NP_001012658.1 | 1 | 129 | ||
| GenBank mRNA | AY761185.1 | - | 1 | 91 | ||
| BC141303.1 | - | 1 | 129 | 9055915 | ||