siRNA Details
| Gene Symbol: Zdhhc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017968.2 | NP_001017968.2 | 4, 5 | 801 | ||
| GenBank mRNA | AY668950.1 | - | 3, 4 | 380 | ||
| BC145714.1 | - | 3, 4 | 432 | |||
| BC145716.1 | - | 3, 4 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc18| Gene Symbol: Zdhhc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017968.2 | NP_001017968.2 | 4, 5 | 801 | ||
| GenBank mRNA | AY668950.1 | - | 3, 4 | 380 | ||
| BC145714.1 | - | 3, 4 | 432 | |||
| BC145716.1 | - | 3, 4 | 432 | |||