siRNA Details
| Gene Symbol: Tex24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013609.2 | NP_001013627.1 | 1 | 844 | ||
| GenBank mRNA | AY552600.1 | - | 1 | 844 | ||
| BC139071.1 | - | 1 | 813 | 8862093 | ||
| BC139072.1 | - | 1 | 813 | 8862094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex24| Gene Symbol: Tex24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013609.2 | NP_001013627.1 | 1 | 844 | ||
| GenBank mRNA | AY552600.1 | - | 1 | 844 | ||
| BC139071.1 | - | 1 | 813 | 8862093 | ||
| BC139072.1 | - | 1 | 813 | 8862094 | ||