siRNA Details
| Gene Symbol: 1190007I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135567.1 | NP_001129039.1 | 3 | 564 | ||
| NM_001135568.1 | NP_001129040.1 | 3 | 435 | |||
| NM_001135569.1 | NP_001129041.1 | 3 | 376 | |||
| GenBank mRNA | AK004515.1 | - | 3 | 426 | ||
| AK045413.1 | - | 3 | 448 | |||
| BC038652.1 | - | 3 | 394 | |||
| BC065417.1 | - | 3 | 398 | |||
| DV649038.1 | - | 3 | 370 | |||

1190007I07Rik
Genomic Map