siRNA Details
| Gene Symbol: 1190007I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135567.1 | NP_001129039.1 | 2 | 359 | ||
| NM_001135568.1 | NP_001129040.1 | 2 | 230 | |||
| NM_001135569.1 | NP_001129041.1 | 2 | 171 | |||
| GenBank mRNA | AK004515.1 | - | 2 | 221 | ||
| AK045413.1 | - | 2 | 243 | |||
| BC038652.1 | - | 2 | 189 | |||
| BC065417.1 | - | 2 | 193 | |||
| DV649038.1 | - | 2 | 165 | |||
| Gene Symbol: 1810014B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007490.1 | - | 2 | 178 | ||
| AK012122.1 | - | 2 | 223 | |||
| AK017634.1 | - | 233 | ||||
| AK019015.1 | - | 2 | 180 | |||
| AK049515.1 | - | 1 | 195 | |||
| AK090016.1 | - | 2 | 220 | |||

1190007I07Rik
Genomic Map