siRNA Details
| Gene Symbol: Gm5782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079904.1 | - | 3 | 505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5782| Gene Symbol: Gm5782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079904.1 | - | 3 | 505 | ||