siRNA Details
| Gene Symbol: Tmem92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034896.2 | NP_001030068.1 | 5 | 527 | ||
| NM_001163172.1 | NP_001156644.1 | 5 | 671 | |||
| GenBank mRNA | AK139848.1 | - | 5 | 527 | ||
| BC147511.1 | - | 5 | 468 | |||
| BC172064.1 | - | 5 | 468 | 9054350 | ||
| BC172068.1 | - | 5 | 412 | |||
| Gene Symbol: Gm11544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205037.1 | NP_001191966.1 | 5 | 665 | ||

Tmem92
Genomic Map