siRNA Details
| Gene Symbol: RING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002931.3 | NP_002922.2 | 3 | 369 | ||
| GenBank mRNA | AK289355.1 | - | 3 | 378 | ||
| AK301010.1 | - | 3 | 378 | |||
| AK301378.1 | - | 3 | 372 | |||
| BC002922.2 | - | 3 | 364 | 3954313 | ||
| BC051866.2 | - | 3 | 362 | 6142438 | ||
| BI259576.1 | - | 3 | 378 | |||
| BT007352.1 | - | 1 | 88 | |||
| Z14000.1 | - | 1 | 163 | |||

RING1
Genomic Map