siRNA Details
| Gene Symbol: Fam181a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195726.1 | NP_001182655.1 | 2 | 753 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam181a| Gene Symbol: Fam181a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195726.1 | NP_001182655.1 | 2 | 753 | ||